Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111256.2 | NP_001104726.1 | 7 | 1045 | 92 | |
GenBank mRNA | AB259650.1 | - | 7 | 1046 | 92 | |
AK346430.1 | - | 7 | 1044 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |