Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112685.2 | NP_001106155.1 | 2 | 349 | 74 | |
XM_005664042.3 | XP_005664099.1 | 1323 | 74 | |||
XM_005664043.3 | XP_005664100.1 | 580 | 74 | |||
XM_005664046.3 | XP_005664103.1 | 639 | 74 | |||
XM_021090863.1 | XP_020946522.1 | 515 | 74 | |||
XM_021090864.1 | XP_020946523.1 | 563 | 74 | |||
XM_021090865.1 | XP_020946524.1 | 627 | 74 | |||
XM_021090866.1 | XP_020946525.1 | 836 | 74 | |||
XM_021090867.1 | XP_020946526.1 | 694 | 74 | |||
GenBank mRNA | AB304400.1 | - | 2 | 351 | 74 | |
AK390264.1 | - | 2 | 362 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |