Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113445.1 | NP_001106916.1 | 5 | 951 | 67 | |
GenBank mRNA | AK232863.1 | - | 5 | 854 | 67 | |
AK232989.1 | - | 5 | 1034 | 67 | ||
AK346145.1 | - | 4 | 911 | 67 | ||
AK393143.1 | - | 5 | 1071 | 67 | ||
AK393199.1 | - | 5 | 1214 | 67 | ||
AK394264.1 | - | 6 | 918 | 67 | ||
AK394277.1 | - | 4 | 912 | 67 | ||
AK394366.1 | - | 5 | 857 | 67 | ||
AK400515.1 | - | 5 | 1218 | 67 | ||
AK400519.1 | - | 5 | 1027 | 67 | ||
EU295557.1 | - | 5 | 951 | 67 | ||
EU717834.1 | - | 5 | 1028 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |