Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123101.2 | NP_001116573.2 | 8 | 1506 | 64 | |
XM_005673332.2 | XP_005673389.1 | 1469 | 64 | |||
XM_005673333.3 | XP_005673390.1 | 1862 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |