Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123135.1 | NP_001116607.1 | 4 | 449 | 72 | |
XM_021076779.1 | XP_020932438.1 | 565 | 72 | |||
XM_021076783.1 | XP_020932442.1 | 648 | 72 | |||
GenBank mRNA | AK400526.1 | - | 5 | 532 | 72 | |
EU421951.1 | - | 4 | 449 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |