Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044537.1 | NP_001038002.1 | 6 - 7 | 710 | 132 | |
GenBank mRNA | DQ325522.1 | - | 6 - 7 | 710 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |