Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044613.1 | NP_001038078.1 | 4 - 5 | 551 | 75 | |
XM_005672836.3 | XP_005672893.1 | 588 | 75 | |||
XM_021077333.1 | XP_020932992.1 | 572 | 75 | |||
XM_021077334.1 | XP_020932993.1 | 533 | 75 | |||
GenBank mRNA | AK239184.1 | - | 4 - 5 | 542 | 75 | |
AY609993.1 | - | 5 - 6 | 496 | 75 | ||
DQ225363.1 | - | 4 - 5 | 551 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |