Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101825.1 | NP_001095295.1 | 4 - 5 | 376 | 81 | |
XM_005665804.3 | XP_005665861.1 | 407 | 81 | |||
GenBank mRNA | AK233579.1 | - | 4 - 5 | 361 | 81 | |
AK236334.1 | - | 4 - 5 | 500 | 81 | ||
AK389553.1 | - | 4 - 5 | 386 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |