Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104956.1 | NP_001098426.1 | 1 - 2 | 211 | 70 | |
XM_021088438.1 | XP_020944097.1 | 293 | 70 | |||
GenBank mRNA | AB081454.1 | - | 1 - 2 | 129 | 70 | |
AB086377.1 | - | 1 - 2 | 168 | 70 | ||
AB267811.1 | - | 1 - 2 | 211 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |