Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105298.1 | NP_001098768.1 | 3 - 4 | 620 | 74 | |
GenBank mRNA | AK233486.1 | - | 3 - 4 | 681 | 74 | |
EF101283.1 | - | 3 - 4 | 620 | 74 | ||
EF101284.1 | - | 3 - 4 | 620 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |