Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109946.1 | NP_001103416.1 | 2 - 3 | 1007 | 63 | |
XM_005660427.3 | XP_005660484.1 | 1553 | 63 | |||
GenBank mRNA | EF154831.1 | - | 2 - 3 | 1007 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |