Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114282.1 | NP_001107754.1 | 3 - 4 | 267 | 121 | |
XM_021082593.1 | XP_020938252.1 | 404 | 121 | |||
GenBank mRNA | AB332237.1 | - | 3 - 4 | 267 | 121 | |
AK239528.1 | - | 3 - 4 | 389 | 121 | ||
AK344997.1 | - | 3 - 4 | 414 | 121 | ||
AK394943.1 | - | 3 - 4 | 389 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |