Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114282.1 | NP_001107754.1 | 4 - 5 | 433 | 113 | |
XM_021082593.1 | XP_020938252.1 | 570 | 113 | |||
GenBank mRNA | AB332237.1 | - | 4 - 5 | 433 | 113 | |
AK239528.1 | - | 4 - 5 | 555 | 113 | ||
AK344997.1 | - | 4 - 5 | 580 | 113 | ||
AK394943.1 | - | 4 - 5 | 555 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |