Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114282.1 | NP_001107754.1 | 6 - 7 | 827 | 120 | |
XM_021082593.1 | XP_020938252.1 | 964 | 120 | |||
GenBank mRNA | AB332237.1 | - | 6 - 7 | 827 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |