Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123156.2 | NP_001116628.1 | 7 - 8 | 925 | 127 | |
XM_021071780.1 | XP_020927439.1 | 1044 | 127 | |||
XM_021071781.1 | XP_020927440.1 | 943 | 127 | |||
GenBank mRNA | AK391517.1 | - | 7 - 8 | 926 | 127 | |
EU402600.2 | - | 7 - 8 | 925 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |