Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNXB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123204.1 | NP_001116676.1 | 4850 | 64 | ||
XM_021098327.1 | XP_020953986.1 | 5615 | 64 | |||
XM_021098328.1 | XP_020953987.1 | 5615 | 64 | |||
XM_021098329.1 | XP_020953988.1 | 5615 | 64 | |||
XM_021098330.1 | XP_020953989.1 | 5615 | 64 | |||
XM_021098331.1 | XP_020953990.1 | 5615 | 64 | |||
XM_021098332.1 | XP_020953991.1 | 5615 | 64 | |||
XM_021098333.1 | XP_020953992.1 | 5615 | 64 | |||
XM_021098334.1 | XP_020953993.1 | 4853 | 64 | |||
XM_021098335.1 | XP_020953994.1 | 5615 | 64 | |||
XM_021098336.1 | XP_020953995.1 | 5615 | 64 | |||
XM_021098337.1 | XP_020953996.1 | 5615 | 64 | |||
XM_021098338.1 | XP_020953997.1 | 5615 | 64 | |||
XM_021098340.1 | XP_020953999.1 | 4853 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa |