Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HADHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348972.1 | NP_001335901.1 | 9 - 10 | 949 | 79 | |
GenBank mRNA | AF028007.1 | - | 6 - 7 | 713 | 79 | |
AK239646.1 | - | 9 - 10 | 896 | 79 | ||
AK343679.1 | - | 9 - 10 | 909 | 79 | ||
AK389670.1 | - | 9 - 10 | 917 | 79 | ||
AK393956.1 | - | 9 - 10 | 885 | 79 | ||
AK400872.1 | - | 9 - 10 | 870 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |