Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP91-PHOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214043.2 | NP_999208.1 | 4 - 5 | 550 | 66 | |
GenBank mRNA | AK230821.1 | - | 7 - 8 | 881 | 66 | |
AK231643.1 | - | 7 - 8 | 882 | 66 | ||
AK232201.1 | - | 7 - 8 | 886 | 66 | ||
AK345180.1 | - | 7 - 8 | 882 | 66 | ||
AK398894.1 | - | 7 - 8 | 861 | 66 | ||
U02476.1 | - | 4 - 5 | 550 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |