Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214083.2 | NP_999248.2 | 2 - 3 | 269 | 70 | |
XM_021079496.1 | XP_020935155.1 | 731 | 70 | |||
XM_021079497.1 | XP_020935156.1 | 541 | 70 | |||
GenBank mRNA | AB040442.1 | - | 2 - 3 | 269 | 70 | |
AK239062.1 | - | 2 - 3 | 302 | 70 | ||
AK390911.1 | - | 2 - 3 | 306 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |