Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001862.1 | NP_001001862.1 | 2 - 3 | 203 | 70 | |
GenBank mRNA | AK347261.1 | - | 3 - 4 | 291 | 70 | |
AY575058.1 | - | 2 - 3 | 203 | 70 | ||
EF673726.1 | - | 3 - 4 | 250 | 70 | ||
EU131524.1 | - | 3 - 4 | 262 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |