Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004040.1 | NP_001004040.1 | 1 - 2 | 161 | 51 | |
XM_021077316.1 | XP_020932975.1 | 486 | 51 | |||
XM_021077317.1 | XP_020932976.1 | 484 | 51 | |||
GenBank mRNA | AK392572.1 | - | 2 - 3 | 307 | 51 | |
V01362.1 | - | 161 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |