Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHCY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011727.1 | NP_001011727.1 | 8 - 9 | 1061 | 62 | |
GenBank mRNA | AB527064.1 | - | 8 - 9 | 969 | 62 | |
AK234979.1 | - | 8 - 9 | 995 | 62 | ||
AK235144.1 | - | 6 - 7 | 638 | 62 | ||
AK239495.1 | - | 8 - 9 | 1095 | 62 | ||
AK346509.1 | - | 3 - 4 | 241 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |