Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031798.1 | NP_001026968.1 | 3 - 4 | 414 | 64 | |
XM_013986257.2 | XP_013841711.1 | 611 | 64 | |||
XM_021079566.1 | XP_020935225.1 | 575 | 64 | |||
GenBank mRNA | AK231821.1 | - | 3 - 4 | 576 | 64 | |
AK238272.1 | - | 3 - 4 | 592 | 64 | ||
AK392729.1 | - | 3 - 4 | 570 | 64 | ||
AY996808.1 | - | 3 - 4 | 414 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |