Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033013.2 | NP_001028185.1 | 3 - 4 | 489 | 75 | |
XM_005663157.3 | XP_005663214.1 | 500 | 75 | |||
XM_005663158.3 | XP_005663215.1 | 498 | 75 | |||
XM_005663159.3 | XP_005663216.1 | 497 | 75 | |||
XM_013996818.2 | XP_013852272.1 | 501 | 75 | |||
XM_013996819.2 | XP_013852273.1 | 500 | 75 | |||
XM_021088750.1 | XP_020944409.1 | 499 | 75 | |||
GenBank mRNA | AK239470.1 | - | 3 - 4 | 488 | 75 | |
AK391270.1 | - | 3 - 4 | 494 | 75 | ||
DQ026064.1 | - | 3 - 4 | 492 | 75 | ||
DQ027013.1 | - | 3 - 4 | 492 | 75 | ||
FJ608551.1 | - | 3 - 4 | 394 | 75 | ||
GQ994431.1 | - | 3 - 4 | 414 | 75 | ||
HM803103.1 | - | 3 - 4 | 417 | 75 | ||
HM803104.1 | - | 3 - 4 | 420 | 75 | ||
KF595284.1 | - | 3 - 4 | 396 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |