Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040640.1 | NP_001035730.1 | 3 - 4 | 399 | 76 | |
XM_021098346.1 | XP_020954005.1 | 2270 | 76 | |||
GenBank mRNA | AY610077.1 | - | 3 - 4 | 432 | 76 | |
DQ329240.2 | - | 3 - 4 | 399 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |