Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213764.1 | NP_998929.1 | 1 | 151 | 65 | |
GenBank mRNA | AK231235.1 | - | 1 | 175 | 65 | |
AK231459.1 | - | 1 | 176 | 65 | ||
AK232665.1 | - | 1 | 175 | 65 | ||
AK233839.1 | - | 1 | 175 | 65 | ||
AK233847.1 | - | 1 | 175 | 65 | ||
AK233892.1 | - | 1 | 175 | 65 | ||
AK233897.1 | - | 1 | 173 | 65 | ||
AK234038.1 | - | 1 | 174 | 65 | ||
AK235379.1 | - | 1 | 173 | 65 | ||
AK236762.1 | - | 1 | 177 | 65 | ||
AK237040.1 | - | 1 | 65 | |||
AK237168.1 | - | 1 | 65 | |||
AK237449.1 | - | 1 | 65 | |||
AK237540.1 | - | 1 | 65 | |||
AK238518.1 | - | 1 | 65 | |||
AK238565.1 | - | 1 | 65 | |||
AK238585.1 | - | 1 | 65 | |||
AK344827.1 | - | 2 | 176 | 65 | ||
AK344886.1 | - | 1 | 173 | 65 | ||
AK345047.1 | - | 1 | 175 | 65 | ||
AK345066.1 | - | 1 | 175 | 65 | ||
AK345079.1 | - | 1 | 175 | 65 | ||
AK346470.1 | - | 1 | 178 | 65 | ||
AK346630.1 | - | 1 | 173 | 65 | ||
AK346711.1 | - | 1 | 175 | 65 | ||
AK346939.1 | - | 1 | 177 | 65 | ||
AK348705.1 | - | 1 | 175 | 65 | ||
AK350403.1 | - | 1 | 65 | |||
AK350453.1 | - | 1 | 65 | |||
AK351198.1 | - | 1 | 65 | |||
AK352328.1 | - | 1 | 65 | |||
AK399717.1 | - | 2 | 65 | |||
AY461380.1 | - | 1 | 151 | 65 | ||
AY610315.1 | - | 2 | 195 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |