Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213791.2 | NP_998956.1 | 3 - 4 | 280 | 102 | |
XM_005670649.3 | XP_005670706.1 | 450 | 102 | |||
GenBank mRNA | AF513016.1 | - | 3 - 4 | 280 | 102 | |
AJ487671.1 | - | 3 - 4 | 280 | 102 | ||
AK344038.1 | - | 4 - 5 | 321 | 102 | ||
DQ629181.1 | - | 3 - 4 | 217 | 102 | ||
JF829998.1 | - | 4 - 5 | 326 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |