Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN1SW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214090.1 | NP_999255.1 | 3 - 4 | 701 | 92 | |
GenBank mRNA | AY091587.1 | - | 3 - 4 | 701 | 92 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Sus scrofa Probe spans exons |