Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214139.1 | NP_999304.1 | 6 - 7 | 871 | 74 | |
GenBank mRNA | AF274055.1 | - | 6 - 7 | 871 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |