Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214244.1 | NP_999409.1 | 1 - 2 | 69 | 67 | |
GenBank mRNA | AF076222.1 | - | 1 - 2 | 69 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |