Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214244.1 | NP_999409.1 | 9 - 10 | 1078 | 68 | |
XM_005673165.3 | XP_005673222.1 | 1615 | 68 | |||
GenBank mRNA | AF076222.1 | - | 9 - 10 | 1078 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |