Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214358.1 | NP_999523.1 | 2 - 3 | 152 | 94 | |
GenBank mRNA | AK230590.1 | - | 2 - 3 | 288 | 94 | |
AK240420.1 | - | 2 - 3 | 320 | 94 | ||
AK392196.1 | - | 2 - 3 | 340 | 94 | ||
AK399677.1 | - | 2 - 3 | 290 | 94 | ||
AY610311.1 | - | 2 - 3 | 271 | 94 | ||
U57333.1 | - | 2 - 3 | 152 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |