Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214441.1 | NP_999606.1 | 1 - 2 | 1450 | 80 | |
GenBank mRNA | AK231433.1 | - | 2 - 3 | 1037 | 80 | |
AY497543.1 | - | 1 - 2 | 1450 | 80 | ||
EU925133.1 | - | 1 - 2 | 960 | 80 | ||
EU925134.1 | - | 1 - 2 | 960 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |