Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128449.1 | NP_001121921.1 | 1 - 2 | 280 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |