Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129961.1 | NP_001123433.1 | 3 - 4 | 355 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |