Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130972.1 | NP_001124444.1 | 1 - 2 | 436 | 77 | |
XM_021089600.1 | XP_020945259.1 | 713 | 77 | |||
GenBank mRNA | EU835903.1 | - | 1 - 2 | 436 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |