Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134345.1 | NP_001127817.1 | 1 - 2 | 69 | 65 | |
GenBank mRNA | EU743945.1 | - | 1 - 2 | 69 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Inventoried Probe spans exons |