Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134345.1 | NP_001127817.1 | 2 - 3 | 186 | 88 | |
GenBank mRNA | EU743945.1 | - | 2 - 3 | 186 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |