Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144847.1 | NP_001138319.1 | 1 | 47 | 70 | |
XM_005662419.3 | XP_005662476.1 | 98 | 70 | |||
XM_021085605.1 | XP_020941264.1 | 46 | 70 | |||
GenBank mRNA | AK231395.1 | - | 2 | 76 | 70 | |
AK344943.1 | - | 2 | 76 | 70 | ||
AK392978.1 | - | 2 | 70 | |||
FJ531495.1 | - | 1 | 47 | 70 | ||
KC250015.1 | - | 1 | 47 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |