Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164512.2 | NP_001157984.1 | 7 - 8 | 1052 | 81 | |
GenBank mRNA | AK234146.1 | - | 7 - 8 | 1011 | 81 | |
AK239514.1 | - | 7 - 8 | 982 | 81 | ||
AK346660.1 | - | 7 - 8 | 982 | 81 | ||
AK390363.1 | - | 7 - 8 | 1052 | 81 | ||
AK392459.1 | - | 7 - 8 | 983 | 81 | ||
AK398777.1 | - | 7 - 8 | 1023 | 81 | ||
AK398909.1 | - | 7 - 8 | 1008 | 81 | ||
GQ368242.1 | - | 6 - 7 | 923 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |