Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144120.1 | NP_001137592.1 | 9 - 10 | 886 | 74 | |
XM_005664661.3 | XP_005664718.2 | 1007 | 74 | |||
GenBank mRNA | AK235121.1 | - | 10 - 11 | 1066 | 74 | |
EU194251.1 | - | 9 - 10 | 886 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |