Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145222.2 | NP_001138694.1 | 2 - 3 | 304 | 81 | |
GenBank mRNA | AY609848.1 | - | 2 - 3 | 311 | 81 | |
EF208119.1 | - | 2 - 3 | 305 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |