Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159615.1 | NP_001153087.1 | 2 - 3 | 617 | 79 | |
GenBank mRNA | AK350973.1 | - | 2 - 3 | 622 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |