Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160087.1 | NP_001153559.1 | 2 - 3 | 748 | 73 | |
XM_021098154.1 | XP_020953813.1 | 878 | 73 | |||
XM_021098155.1 | XP_020953814.1 | 1178 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |