Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161640.1 | NP_001155112.1 | 5 - 6 | 964 | 68 | |
XM_021074526.1 | XP_020930185.1 | 983 | 68 | |||
GenBank mRNA | AB499815.1 | - | 5 - 6 | 964 | 68 | |
AK232925.1 | - | 5 - 6 | 920 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |