Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162888.1 | NP_001156360.1 | 5 - 6 | 529 | 100 | |
XM_021072043.1 | XP_020927702.1 | 672 | 100 | |||
XM_021072044.1 | XP_020927703.1 | 657 | 100 | |||
XM_021072045.1 | XP_020927704.1 | 703 | 100 | |||
GenBank mRNA | AB506709.1 | - | 5 - 6 | 529 | 100 | |
AK349916.1 | - | 6 - 7 | 625 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons |