Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171751.1 | NP_001165222.1 | 7 - 8 | 1159 | 72 | |
XM_005655517.3 | XP_005655574.2 | 1420 | 72 | |||
XM_005655519.3 | XP_005655576.2 | 1044 | 72 | |||
XM_013988213.2 | XP_013843667.2 | 1036 | 72 | |||
GenBank mRNA | AK343272.1 | - | 7 - 8 | 1258 | 72 | |
AY610084.1 | - | 7 - 8 | 1190 | 72 | ||
GQ369460.1 | - | 7 - 8 | 1159 | 72 | ||
GQ369461.1 | - | 7 - 8 | 1159 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |