Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177924.1 | NP_001171395.1 | 4 - 5 | 469 | 90 | |
GenBank mRNA | GU475001.1 | - | 4 - 5 | 469 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons |