Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184753.1 | NP_001171682.1 | 7 - 8 | 723 | 59 | |
XM_021063977.1 | XP_020919636.1 | 789 | 59 | |||
GenBank mRNA | GU475009.1 | - | 7 - 8 | 723 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |