Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185049.2 | NP_001171978.2 | 2 - 3 | 2176 | 114 | |
XM_021100320.1 | XP_020955979.1 | 2176 | 114 | |||
XM_021100326.1 | XP_020955985.1 | 594 | 114 | |||
GenBank mRNA | HM037345.1 | - | 1 - 2 | 232 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |